Learn More
Abcam N6-Cyclopentyladenosine (CPA), A1 agonist, 25MG
Catalog No. 502707609
Encompass_Preferred
MW 335.36 Da, Purity >99%. Potent and selective adenosine A1 receptor agonist (Ki values are 2.3, 790 and 43 nM for human A1, A2A and A3 receptors, respectively). Centrally active following systemic administration *in vivo*. Anticonvulsant, with cardiovacular actions, and effects on sleep regulation.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7609 Supplier Abcam Supplier No. AB12039825MG
MW 335.36 Da, Purity >99%. Potent and selective adenosine A1 receptor agonist (Ki values are 2.3, 790 and 43 nM for human A1, A2A and A3 receptors, respectively). Centrally active following systemic administration *in vivo*. Anticonvulsant, with cardiovacular actions, and effects on sleep regulation.
The product is subject to the following: Abcam Restricted Use Statement