Learn More
Medchemexpress LLC HY-111646 5mg Medchemexpress, N6-Etheno 2'-deoxyadenosine CAS:68498-25-9 Purity:>98%
SDP
Catalog No. 502080800
Encompass_Preferred
Medchemexpress, HY-111646 5mg N6-Etheno 2'-deoxyadenosine CAS:68498-25-9 N6-Etheno 2'-deoxyadenosine is a reactive oxygen species (ROS)/reactive nitrogen species (RNS)-induced DNA oxidation product, used as a biomarker to evaluate chronic inflammation and lipid peroxidation in animal or human tissues. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0800 Supplier Medchemexpress LLC Supplier No. HY1116465MG
Medchemexpress, HY-111646 5mg N6-Etheno 2'-deoxyadenosine CAS:68498-25-9 N6-Etheno 2'-deoxyadenosine is a reactive oxygen species (ROS)/reactive nitrogen species (RNS)-induced DNA oxidation product, used as a biomarker to evaluate chronic inflammation and lipid peroxidation in animal or human tissues. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.