Learn More
Medchemexpress LLC N6-Isopentenyladenosine | 7724-76-7 | 99.4% | 335.36 | 1 ML
SDP
Catalog No. 5000453619
Encompass_Preferred
N6-Isopentenyladenosine (Riboprine) is an RNA modification found in cytokinins, which are known to regulate plant growth and differentiation. It is also present in a subset of tRNAs, where it enhances the efficiency and accuracy of translation. This compound is an end product of the mevalonate pathway and functions as an autophagy inhibitor, demonstrating interesting anti-melanoma activity.
  • RNA modification found in cytokinins
  • Regulates plant growth/differentiation
  • Improves efficiency and accuracy of translation in tRNAs
  • End product of the mevalonate pathway
  • Autophagy inhibitor
  • Exhibits anti-melanoma activity
Catalog No. 50-004-53619 Supplier Medchemexpress LLC Supplier No. HYW01120910MM/1ML
N6-Isopentenyladenosine (Riboprine) is an RNA modification found in cytokinins, which are known to regulate plant growth and differentiation. It is also present in a subset of tRNAs, where it enhances the efficiency and accuracy of translation. This compound is an end product of the mevalonate pathway and functions as an autophagy inhibitor, demonstrating interesting anti-melanoma activity.
  • RNA modification found in cytokinins
  • Regulates plant growth/differentiation
  • Improves efficiency and accuracy of translation in tRNAs
  • End product of the mevalonate pathway
  • Autophagy inhibitor
  • Exhibits anti-melanoma activity