Learn More
Cayman Chemical N6-Methyl-2-deoxyadenosIn 25mg
SDP
Catalog No. 502944104
Encompass_Preferred
A nucleoside and inducer of EPC proliferation; stimulates the proliferation of EPCs isolated from mouse fetal livers and colony formation of isolated mouse bone marrow cell-derived progenitor cells at 10 and 100 µM; accumulates along promoters and gene coding sequences in activated mouse prefrontal cortical neurons; is associated with activation at the Bdnf promoter
Catalog No. 50-294-4104 Supplier Cayman Chemical Supplier No. 4040725 MG
A nucleoside and inducer of EPC proliferation; stimulates the proliferation of EPCs isolated from mouse fetal livers and colony formation of isolated mouse bone marrow cell-derived progenitor cells at 10 and 100 µM; accumulates along promoters and gene coding sequences in activated mouse prefrontal cortical neurons; is associated with activation at the Bdnf promoter