Learn More
Apexbio Technology LLC N6-Methyl-ATP 50ul (100 mM)
SDP
Catalog No. 5000062768
Encompass_Preferred
N6-Methyl-ATP is a methylated derivative of adenosine triphosphate (ATP) designed to function as a selective agonist for purinergic P2 receptors Due to structural modifications including methylation at the N6 position of the adenine moiety this compound displays specificity for interaction with distinct subtypes of P2 purinergic receptors In cellular and molecular research N6-Methyl-ATP is utilized as a pharmacological tool to investigate receptor-mediated signaling pathways particularly ion channel activation calcium influx and associated intracellular signaling cascades modulated through P2 receptors Furthermore it serves as a molecular probe in characterizing purinergic receptor-mediated neurotransmission physiological regulatory processes and receptor subtype distribution in both normal physiological states and pathological conditions contributing to the foundational understanding necessary for receptor-targeted drug discovery
Catalog No. 50-000-62768 Supplier Apexbio Technology LLC Supplier No. B79660.05
N6-Methyl-ATP is a methylated derivative of adenosine triphosphate (ATP) designed to function as a selective agonist for purinergic P2 receptors Due to structural modifications including methylation at the N6 position of the adenine moiety this compound displays specificity for interaction with distinct subtypes of P2 purinergic receptors In cellular and molecular research N6-Methyl-ATP is utilized as a pharmacological tool to investigate receptor-mediated signaling pathways particularly ion channel activation calcium influx and associated intracellular signaling cascades modulated through P2 receptors Furthermore it serves as a molecular probe in characterizing purinergic receptor-mediated neurotransmission physiological regulatory processes and receptor subtype distribution in both normal physiological states and pathological conditions contributing to the foundational understanding necessary for receptor-targeted drug discovery