Learn More
Medchemexpress LLC N6-Methyl-dA phosphoramidite | 105931-58-6 | 99.8% | 767.85 | 100 MG
SDP
Catalog No. 5000350768
Encompass_Preferred
N6-Methyl-dA phosphoramidite is an active compound used in the synthesis of oligodeoxyribonucleotides. It is intended for research use only and is not for sale to patients. The product is available as a solid, white to off-white in color, and has a molecular weight of 767.85.
  • Utilized in the synthesis of oligodeoxyribonucleotides.
  • High purity of 99.83% ensures reliable experimental results.
  • Product supplied in solid form, white to off-white in color.
  • Can be stored at 4°C in sealed storage, away from moisture and light.
  • Stock solutions in solvent can be stored at -80°C for 6 months or -20°C for 1 month.
  • Available in various quantities, including milligrams and grams.
  • Detailed technical support and documentation available, including data sheet, COA, and SDS.
  • Soluble in DMSO at 250 mg/mL, requiring sonication.
Catalog No. 50-003-50768 Supplier Medchemexpress LLC Supplier No. HY138582100MG
N6-Methyl-dA phosphoramidite is an active compound used in the synthesis of oligodeoxyribonucleotides. It is intended for research use only and is not for sale to patients. The product is available as a solid, white to off-white in color, and has a molecular weight of 767.85.
  • Utilized in the synthesis of oligodeoxyribonucleotides.
  • High purity of 99.83% ensures reliable experimental results.
  • Product supplied in solid form, white to off-white in color.
  • Can be stored at 4°C in sealed storage, away from moisture and light.
  • Stock solutions in solvent can be stored at -80°C for 6 months or -20°C for 1 month.
  • Available in various quantities, including milligrams and grams.
  • Detailed technical support and documentation available, including data sheet, COA, and SDS.
  • Soluble in DMSO at 250 mg/mL, requiring sonication.