Learn More
Cayman Chemical N6-Methyladenine, Purity: ≥98%, CAS: 443-72-1, 100 mg
SDP
Catalog No. 502979823
Encompass_Preferred
A modified purine that is commonly found in genomes of prokaryotes, protists, and plants
Catalog No. 50-297-9823 Supplier Cayman Chemical Supplier No. 18153100 MG
A modified purine that is commonly found in genomes of prokaryotes, protists, and plants