Learn More
Cayman Chemical N6-Methyladenine, Purity: ≥98%, CAS: 443-72-1, 250 mg
SDP
Catalog No. 502980511
Encompass_Preferred
A modified purine that is commonly found in genomes of prokaryotes, protists, and plants
Catalog No. 50-298-0511 Supplier Cayman Chemical Supplier No. 18153250 MG
A modified purine that is commonly found in genomes of prokaryotes, protists, and plants