Learn More
Cayman Chemical N6-MethyladenIn 500mg
SDP
Catalog No. 502979461
Encompass_Preferred
A modified purine that is commonly found in genomes of prokaryotes, protists, and plants
Catalog No. 50-297-9461 Supplier Cayman Chemical Supplier No. 18153500 MG
A modified purine that is commonly found in genomes of prokaryotes, protists, and plants