Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N6-Methyladenine 10mM 1mL | 443-72-1 | 1 ML
SDP

Catalog No. 5000226213
Encompass_Preferred

N6-methyladenine is a methylated form of the nucleobase adenine used as an analytical standard and research reagent for studies of DNA/RNA methylation and bacterial epigenetics. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and is also available as solids for reconstitution.

  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • Also available as solid material for reconstitution.
  • High purity (98.8%).
  • Molecular formula C6H7N5 and molecular weight 149.16 g/mol.
  • Intended for research and analytical applications.

Catalog No. 50-002-26213 Supplier Medchemexpress LLC Supplier No. HY11688710MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N6-methyladenine is a methylated form of the nucleobase adenine used as an analytical standard and research reagent for studies of DNA/RNA methylation and bacterial epigenetics. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and is also available as solids for reconstitution.

  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • Also available as solid material for reconstitution.
  • High purity (98.8%).
  • Molecular formula C6H7N5 and molecular weight 149.16 g/mol.
  • Intended for research and analytical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.