Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N6-Methyladenine 250mg | 443-72-1 | 149.16 g/mol | C6H7N5 | 250 MG
SDP

Catalog No. 5000226214
Encompass_Preferred

N6-Methyladenine is a methylated purine nucleobase provided as an analytical standard for research and analytical applications. Supplied as a 250 mg solid, it is intended for use in epigenetics, microbiology, and analytical method development where a high-purity reference compound is required.

  • Purity 98.79% (typical)
  • Molecular formula C6H7N5; molecular weight 149.16 g/mol
  • Appearance: solid, supplied as 250 mg
  • Solubility: DMSO 20 mg/mL; ethanol 2 mg/mL with warming
  • Storage: 4°C, protect from light; in solvent -80°C (up to 6 months)

Catalog No. 50-002-26214 Supplier Medchemexpress LLC Supplier No. HY116887250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N6-Methyladenine is a methylated purine nucleobase provided as an analytical standard for research and analytical applications. Supplied as a 250 mg solid, it is intended for use in epigenetics, microbiology, and analytical method development where a high-purity reference compound is required.

  • Purity 98.79% (typical)
  • Molecular formula C6H7N5; molecular weight 149.16 g/mol
  • Appearance: solid, supplied as 250 mg
  • Solubility: DMSO 20 mg/mL; ethanol 2 mg/mL with warming
  • Storage: 4°C, protect from light; in solvent -80°C (up to 6 months)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.