Learn More
Cayman Chemical N6-Methyladenosine, Purity: ≥95%, CAS: 1867-73-8, 1 g
SDP
Catalog No. 502939080
Encompass_Preferred
An adenosine analog; inhibits epinephrine-induced contraction of isolated guinea pig ileum and thoracic aorta at 10 μM; decreases arterial blood pressure and renal blood flow and increases peripheral resistance in anesthetized dogs at 1 mg/kg; inhibits tumor growth in the C3H/ST and C3HB/ST mouse models of spontaneous mammary adenocarcinomas; the most prevelant mRNA modification in eukaryotes and has roles in cell viability and development
Catalog No. 50-293-9080 Supplier Cayman Chemical Supplier No. 288331 G
An adenosine analog; inhibits epinephrine-induced contraction of isolated guinea pig ileum and thoracic aorta at 10 μM; decreases arterial blood pressure and renal blood flow and increases peripheral resistance in anesthetized dogs at 1 mg/kg; inhibits tumor growth in the C3H/ST and C3HB/ST mouse models of spontaneous mammary adenocarcinomas; the most prevelant mRNA modification in eukaryotes and has roles in cell viability and development