Learn More
Cayman Chemical N6-MethyladenosIn 500mg
SDP
Catalog No. 502939058
Encompass_Preferred
An adenosine analog; inhibits epinephrine-induced contraction of isolated guinea pig ileum and thoracic aorta at 10 μM; decreases arterial blood pressure and renal blood flow and increases peripheral resistance in anesthetized dogs at 1 mg/kg; inhibits tumor growth in the C3H/ST and C3HB/ST mouse models of spontaneous mammary adenocarcinomas; the most prevelant mRNA modification in eukaryotes and has roles in cell viability and development
Catalog No. 50-293-9058 Supplier Cayman Chemical Supplier No. 28833500 MG
An adenosine analog; inhibits epinephrine-induced contraction of isolated guinea pig ileum and thoracic aorta at 10 μM; decreases arterial blood pressure and renal blood flow and increases peripheral resistance in anesthetized dogs at 1 mg/kg; inhibits tumor growth in the C3H/ST and C3HB/ST mouse models of spontaneous mammary adenocarcinomas; the most prevelant mRNA modification in eukaryotes and has roles in cell viability and development