Learn More
Medchemexpress LLC HY-N0086 100mg , N6-Methyladenosine CAS:1867-73-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0086100MG
Encompass_Preferred
HY-N0086 100mg N6-Methyladenosine CAS: 1867-73-8 N6-Methyladenosine is the most prevalent internal (non-cap) modification present in the messenger RNA (mRNA) of all higher eukaryotes. N6-Methyladenosine can modifies viral RNAs and has antiviral activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1543