Learn More
Trilink Bio Technologies Inc N6-Methyladenosine-5'-Triphosphate, 10 µmole
SDP
Catalog No. NC0889204
Encompass
Molecular Formula: C11H18N5O13P3, Purity Specification: ≥90% by AX-HPLC, Molecular Weight: 521.2 g/mol, Salt Form: Lithium
Catalog No. NC0889204 Supplier Trilink Bio Technologies Inc Supplier No. N101310
Add to Cart
Add to Cart
Molecular Formula: C11H18N5O13P3, Purity Specification: ≥90% by AX-HPLC, Molecular Weight: 521.2 g/mol, Salt Form: Lithium