Learn More
Selleck Chemical LLC N6-methyladenosine (m6A) 50mg 1867-73-8 NSC-29409
SDP
Supplier:  Selleck Chemical LLC S319050MG
Encompass_Preferred
N6-methyladenosine (m6A, NSC-29409) is a base modified analog of adenosine and is found as a minor nucleoside in natural RNAs. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3483