Learn More
N6-methyladenosine (m6A) Rabbit anti-Human, Clone: RM362, Novus Biologicals™

Description
Specifications
Specifications
| Antigen | N6-methyladenosine (m6A) |
| Applications | Dot Blot, ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | RM362 |
| Conjugate | Unconjugated |
| Dilution | ELISA 1:100 - 1:2000, Immunoprecipitation, Dot Blot 1:100 - 1:2000 |
| Formulation | 50% Glycerol/PBS with 1% BSA |
| Gene Alias | (-)-6-(Methylamino)purine riboside, N(6)-Methyladenosine, N-Methyladenosine |
| Host Species | Rabbit |
| Immunogen | BSA-conjugated N6-methyladenosine (m6A) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.