Learn More
Cayman Chemical N6,N6-Dimethyladenosine, Purity: ≥95%, CAS: 2620-62-4, 1 g
SDP
Catalog No. 502913643
Encompass_Preferred
A modified nucleoside; has been found in tRNA isolated from M. bovis; dimethylation of adjacent adenosine nucleosides is found in E. coli 16S rRNA and is important for the initiation of protein synthesis; inhibits the proliferation of L1210 leukemia cells in vitro (IC50 = 0.5 µg/ml) but does not have antitumor activity in vivo; a component of puromycin
Catalog No. 50-291-3643 Supplier Cayman Chemical Supplier No. 353481 G
A modified nucleoside; has been found in tRNA isolated from M. bovis; dimethylation of adjacent adenosine nucleosides is found in E. coli 16S rRNA and is important for the initiation of protein synthesis; inhibits the proliferation of L1210 leukemia cells in vitro (IC50 = 0.5 µg/ml) but does not have antitumor activity in vivo; a component of puromycin