Learn More
Cayman Chemical N6 N6-DImethyladenosIn 500mg
SDP
Catalog No. 502909988
Encompass_Preferred
A modified nucleoside; has been found in tRNA isolated from M. bovis; dimethylation of adjacent adenosine nucleosides is found in E. coli 16S rRNA and is important for the initiation of protein synthesis; inhibits the proliferation of L1210 leukemia cells in vitro (IC50 = 0.5 UG/ml) but does not have antitumor activity in vivo; a component of puromycin
Catalog No. 50-290-9988 Supplier Cayman Chemical Supplier No. 35348500 MG
A modified nucleoside; has been found in tRNA isolated from M. bovis; dimethylation of adjacent adenosine nucleosides is found in E. coli 16S rRNA and is important for the initiation of protein synthesis; inhibits the proliferation of L1210 leukemia cells in vitro (IC50 = 0.5 UG/ml) but does not have antitumor activity in vivo; a component of puromycin