Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N6,N6-dimethyladenosine | 2620-62-4 | MFCD00022822 | 100.0% | 295.29 g·mol⁻1 | C12H17N5O4 | 1 ML
SDP

Catalog No. 5000247239
Encompass_Preferred

N6,N6-dimethyladenosine is a modified ribonucleoside used as an analytical standard and research reagent. It is supplied as solid material or as a 10 mM solution in DMSO and is used in studies of RNA modification, cellular signaling (including AKT pathway modulation), and viral entry protein interactions.

  • High purity (>99.9%) suitable for analytical use.
  • Available as solid and 10 mM solution in DMSO for convenient handling.
  • Molecular formula C12H17N5O4 and molecular weight 295.29 g·mol⁻1.
  • Suitable for studies of RNA modification and cellular signaling.
  • Certificate of analysis and safety data sheet available.

Catalog No. 50-002-47239 Supplier Medchemexpress LLC Supplier No. HY10198410MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N6,N6-dimethyladenosine is a modified ribonucleoside used as an analytical standard and research reagent. It is supplied as solid material or as a 10 mM solution in DMSO and is used in studies of RNA modification, cellular signaling (including AKT pathway modulation), and viral entry protein interactions.

  • High purity (>99.9%) suitable for analytical use.
  • Available as solid and 10 mM solution in DMSO for convenient handling.
  • Molecular formula C12H17N5O4 and molecular weight 295.29 g·mol⁻1.
  • Suitable for studies of RNA modification and cellular signaling.
  • Certificate of analysis and safety data sheet available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.