Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N6-propionyl-L-lysine | 1974-17-0 | MFCD26793597 | 99.9% | 202.25 g/mol | C9H18N2O3 | 100 MG
SDP

Catalog No. 5000209891
Encompass_Preferred

N6-propionyl-L-lysine is an N6-acylated derivative of the amino acid L-lysine supplied as a white to off-white solid for research use. It is commonly used as a biochemical building block and reference standard in peptide modification and epigenetic studies.

  • Chemical formula C9H18N2O3, molecular weight 202.25 g/mol.
  • Purity 99.86% as supplied.
  • Soluble in water at 100 mg/mL; may require ultrasonic agitation.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solution -80°C (6 months) or -20°C (1 month).
  • Available pack sizes include 100 mg, 250 mg, 1 g, 5 g, 10 g, and 50 g.

Catalog No. 50-002-09891 Supplier Medchemexpress LLC Supplier No. HYW048207100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N6-propionyl-L-lysine is an N6-acylated derivative of the amino acid L-lysine supplied as a white to off-white solid for research use. It is commonly used as a biochemical building block and reference standard in peptide modification and epigenetic studies.

  • Chemical formula C9H18N2O3, molecular weight 202.25 g/mol.
  • Purity 99.86% as supplied.
  • Soluble in water at 100 mg/mL; may require ultrasonic agitation.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solution -80°C (6 months) or -20°C (1 month).
  • Available pack sizes include 100 mg, 250 mg, 1 g, 5 g, 10 g, and 50 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.