Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N6-propionyl-L-lysine | 1974-17-0 | MFCD26793597 | 99.9% | 202.25 g/mol | C9H18N2O3 | 250 MG
SDP

Catalog No. 5000209275
Encompass_Preferred

N6-propionyl-L-lysine is a propionylated derivative of L-lysine supplied as a solid for laboratory research and biochemical studies.

  • Molecular formula: C9H18N2O3.
  • Molecular weight: 202.25 g/mol.
  • CAS number: 1974-17-0.
  • Purity typically reported at ~99.9% (manufacturer documents contain close values).
  • Appearance: white to off-white solid.
  • Applications: studies of lysine acylation, epigenetic modification assays, and amino acid derivative research.
  • Available packaging: e.g., 100 mg, 250 mg, 1 g, 5 g, 10 g, 50 g.

Catalog No. 50-002-09275 Supplier Medchemexpress LLC Supplier No. 250MGHYW048207
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N6-propionyl-L-lysine is a propionylated derivative of L-lysine supplied as a solid for laboratory research and biochemical studies.

  • Molecular formula: C9H18N2O3.
  • Molecular weight: 202.25 g/mol.
  • CAS number: 1974-17-0.
  • Purity typically reported at ~99.9% (manufacturer documents contain close values).
  • Appearance: white to off-white solid.
  • Applications: studies of lysine acylation, epigenetic modification assays, and amino acid derivative research.
  • Available packaging: e.g., 100 mg, 250 mg, 1 g, 5 g, 10 g, 50 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.