Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N6-Threonylcarbamoyladenosine | 24719-82-2 | 99.6% | 412.35 | 1 MG
SDP

Catalog No. 5000387834
Encompass_Preferred

N6-Threonylcarbamoyladenosine is a common nucleoside that can decorate and become tRNA. It is for research use only.

  • Purity of 99.62%
  • Molecular weight of 412.35
  • Chemical formula: C15H20N6O8
  • Appearance: Solid
  • Color: Off-white to light brown
  • Storage conditions: -20°C for powder, -80°C for 6 months in solvent

Catalog No. 50-003-87834 Supplier Medchemexpress LLC Supplier No. HY183981MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N6-Threonylcarbamoyladenosine is a common nucleoside that can decorate and become tRNA. It is for research use only.

  • Purity of 99.62%
  • Molecular weight of 412.35
  • Chemical formula: C15H20N6O8
  • Appearance: Solid
  • Color: Off-white to light brown
  • Storage conditions: -20°C for powder, -80°C for 6 months in solvent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.