Learn More
Apexbio Technology LLC N6022(Synonyms: N6022, GSNOR inhibitor N6022, N-6022, N 6022, GSNOR Inhibitor N-6022, CAS 1208315-24-5, GSNORi N6022), 10mM (in 1mL DMSO), CAS: 1208315-24-5.
SDP
Catalog No. 5000060059
Encompass_Preferred
N6022 (CAS 1208315-24-5) is a selective reversible inhibitor of S-nitrosoglutathione reductase (GSNOR) an enzyme involved in regulating the levels of S-nitrosoglutathione (GSNO) Inhibition of GSNOR leads to elevated GSNO concentrations promoting vasodilatory and anti-inflammatory effects In biochemical assays N6022 exhibits IC50 values of 8 nM (GSNO reduction assay) and 32 nM (HMGSH oxidation assay) with corresponding Ki values of 2 5 nM and 3 1 nM respectively Due to its selectivity profile and potency N6022 has entered clinical evaluation for inflammatory pulmonary conditions
Catalog No. 50-000-60059 Supplier Apexbio Technology LLC Supplier No. B11115.1
N6022 (CAS 1208315-24-5) is a selective reversible inhibitor of S-nitrosoglutathione reductase (GSNOR) an enzyme involved in regulating the levels of S-nitrosoglutathione (GSNO) Inhibition of GSNOR leads to elevated GSNO concentrations promoting vasodilatory and anti-inflammatory effects In biochemical assays N6022 exhibits IC50 values of 8 nM (GSNO reduction assay) and 32 nM (HMGSH oxidation assay) with corresponding Ki values of 2 5 nM and 3 1 nM respectively Due to its selectivity profile and potency N6022 has entered clinical evaluation for inflammatory pulmonary conditions