Learn More
Cayman Chemical N6-Benzyladenosine-5'-O-diphosphate (sodium salt), Purity: ≥95%, 2.6 mg
SDP
Catalog No. 502916724
Encompass_Preferred
An ADP derivative and active metabolite of N6-benzyladenine; inhibits the ATPase activities of mortalin, Hsp70, and Hsc70 (apparent Kis = 86.51, 294.5, and 1,612 µM, respectively, for the human enzymes); inhibits the proliferation of CEM, HL-60, K562, RPMI-8226, and MCF-7 cells (IC50s = 2, 1.3, 4, 4.5, and 8 µM, respectively); has been used in the synthesis of radiolabeled and unlabeled forms of N6-benzyl-ATPγS
Catalog No. 50-291-6724 Supplier Cayman Chemical Supplier No. 383702.6 MG
An ADP derivative and active metabolite of N6-benzyladenine; inhibits the ATPase activities of mortalin, Hsp70, and Hsc70 (apparent Kis = 86.51, 294.5, and 1,612 µM, respectively, for the human enzymes); inhibits the proliferation of CEM, HL-60, K562, RPMI-8226, and MCF-7 cells (IC50s = 2, 1.3, 4, 4.5, and 8 µM, respectively); has been used in the synthesis of radiolabeled and unlabeled forms of N6-benzyl-ATPγS