Learn More
Cayman Chemical N6BenzyladenosIn5OtrIph 3mg
SDP
Catalog No. 502916765
Encompass_Preferred
An N-phenyl-substituted analog of ATP and a ligand used for bump and hole; is 7-fold selective for inhibiting the ATP binding and ATPase activity of MLCY61G over the wild-type enzyme; has been used to develop a technique to identify substrates of Adi3 in tomato plants
Catalog No. 50-291-6765 Supplier Cayman Chemical Supplier No. 383713 MG
An N-phenyl-substituted analog of ATP and a ligand used for bump and hole; is 7-fold selective for inhibiting the ATP binding and ATPase activity of MLCY61G over the wild-type enzyme; has been used to develop a technique to identify substrates of Adi3 in tomato plants