Learn More
Selleck Chemical LLC N6F11-E1812-250MG
SDP
Catalog No. 503138931
Encompass_Preferred
N6F11 is an inducer of ferroptosis N6F11 is bound to the RING domain of the E3 ubiquitin ligase tripartite motif containing 25 (TRIM25) in cancer cells to trigger TRIM25-mediated K48-linked ubiquitination of glutathione peroxidase 4 (GPX4) resulting in proteasomal degradation
Catalog No. 50-313-8931 Supplier Selleck Chemical LLC Supplier No. E1812250MG
N6F11 is an inducer of ferroptosis N6F11 is bound to the RING domain of the E3 ubiquitin ligase tripartite motif containing 25 (TRIM25) in cancer cells to trigger TRIM25-mediated K48-linked ubiquitination of glutathione peroxidase 4 (GPX4) resulting in proteasomal degradation