Learn More
Cayman Chemical N6MethyladenosIn 5monpho 10mg
SDP
Catalog No. 502993726
Encompass_Preferred
An activator of glycogen phosphorylase b (Ka = 22 µM); a non-competitive inhibitor of rat adenylate kinase II; has been used for immunoprecipitation of N6-methyladenosine
Catalog No. 50-299-3726 Supplier Cayman Chemical Supplier No. 2338210 MG
An activator of glycogen phosphorylase b (Ka = 22 µM); a non-competitive inhibitor of rat adenylate kinase II; has been used for immunoprecipitation of N6-methyladenosine