Learn More
Medchemexpress LLC N7-methyl-guanosine-5'-triphosphate-5'-adenosine diammonium | 75252-10-7 | C21H35N12O17P3 | 10MG
SDP
Catalog No. 5000261835
Encompass_Preferred
m7G(5')ppp(5')A diammonium is a dinucleotide 5' cap analog that mimics the natural eukaryotic mRNA cap structure. It is used to produce capped RNA in vitro for studies of translation initiation, mRNA stability, and cap-binding protein interactions.
  • Mimics natural 5' mRNA cap for biologically relevant capping.
  • Suitable for in vitro transcription and translation assays.
  • High purity (>99%) supports reproducible experimental results.
  • Solid, white to off-white material simplifies handling and storage.
  • Stable under recommended storage conditions for consistent performance.
Catalog No. 50-002-61835 Supplier Medchemexpress LLC Supplier No. HY139100B10MG
m7G(5')ppp(5')A diammonium is a dinucleotide 5' cap analog that mimics the natural eukaryotic mRNA cap structure. It is used to produce capped RNA in vitro for studies of translation initiation, mRNA stability, and cap-binding protein interactions.
  • Mimics natural 5' mRNA cap for biologically relevant capping.
  • Suitable for in vitro transcription and translation assays.
  • High purity (>99%) supports reproducible experimental results.
  • Solid, white to off-white material simplifies handling and storage.
  • Stable under recommended storage conditions for consistent performance.