Learn More
Cayman Chemical N8-Acetylspermidine (hydrochloride), Purity: ≥95%, CAS: 34450-15-2, 10 mg
SDP
Catalog No. 502917502
Encompass_Preferred
An endogenous polyamine; urinary levels are increased in patients with chronic pancreatitis; blood levels are increased in patients with ischemic cardiomyopathy, as well as in patients with Parkinson’s disease
Catalog No. 50-291-7502 Supplier Cayman Chemical Supplier No. 3743410 MG
An endogenous polyamine; urinary levels are increased in patients with chronic pancreatitis; blood levels are increased in patients with ischemic cardiomyopathy, as well as in patients with Parkinson’s disease