Learn More
Poltex N95 Face Mask with Shield Dispenser - Sliding Door, Front Load, Wall Mount
SDP
Catalog No. 22022539
Change view
Click to view available options
Product Type:
Face Mask Dispenser
Catalog No. Product Type
22-022-539 Face Mask Dispenser
1 options
Catalog No. 22-022-539 Supplier Poltex Supplier No. SHLDN95CABW
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
Front load holder for both a N95 mask box and face mask with shield holder. Fits halyard fluid shield 2 (62114) or similar surgical mask, and two boxes of Fluid shield 3 (46767) N95 mask boxes. Keyhole slots for mounting. Wall mount (screws/anchors included).
  • Front load holder for n95 mask box and face mask with shield holder
  • Fits Halyard fluid shield 2 (62114) or similar surgical mask, and two boxes of fluid shield 3 (46767) N95 mask boxes
  • Super durable PETG construction
  • Mounts to wall with keyhole slots (screws/anchors included)
Specifications
Product Type Face Mask Dispenser