Learn More
Ironwear N95 Respirator | NIOSH Approved | 20/bx
Catalog No. NC1842584
Encompass_Preferred
Our N95 NIOSH certified respirator was designed for a secure fit. Two elastic bands allow for earless placement above and below areas typically bothered by ear-loop designs, making this mask ideal for long term wear. A folded face design creates minimal discomfort and space for the wearer to feel free and able to breathe while reducing nose and mouth irritation. The soft texture of the Ironwear 1511 provides all necessary protections from particles and infectious sprays while meeting filtration requirements with its adjustable nose piece. The compatibility of this mask makes it an ideal fit; compatible with a variety of protective eyewear and other secondary protective gear.
This item is not returnable. View return policy
Catalog No. NC1842584 Supplier Ironwear Supplier No. 1511
Our N95 NIOSH certified respirator was designed for a secure fit. Two elastic bands allow for earless placement above and below areas typically bothered by ear-loop designs, making this mask ideal for long term wear. A folded face design creates minimal discomfort and space for the wearer to feel free and able to breathe while reducing nose and mouth irritation. The soft texture of the Ironwear 1511 provides all necessary protections from particles and infectious sprays while meeting filtration requirements with its adjustable nose piece. The compatibility of this mask makes it an ideal fit; compatible with a variety of protective eyewear and other secondary protective gear.