Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ N95 Particulate Welding Respirators

Catalog No. 19002710 Shop All 3M Products
Change view
Click to view available options
Nuisance Odor Relief:
None
Certifications (NIOSH):
NIOSH 42 CFR 84, N95, approval number: TC-84A-0454
NIOSH 42 CFR 84, N95, approval number: TC-84A-1299
Strap Attachment Type:
Adjustable Buckle Straps
Braided Headbands
With Faceseal:
No
Yes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Nuisance Odor Relief Certifications (NIOSH) Strap Attachment Type With Faceseal
19-002-710 None NIOSH 42 CFR 84, N95, approval number: TC-84A-0454 Adjustable Buckle Straps Yes
19-038-989 None NIOSH 42 CFR 84, N95, approval number: TC-84A-1299 Braided Headbands No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Do not exceed maximum use concentration established by regulatory standards. If respirator becomes damaged, soiled, or breathing becomes difficult, leave the contaminated area and dispose of the respirator. Before each use of the product, please inspect for any rips, damage, tears, etc. These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Catalog No. 19-002-710 Supplier 3M™ Supplier No. 7000002027
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Do not exceed maximum use concentration established by regulatory standards. If respirator becomes damaged, soiled, or breathing becomes difficult, leave the contaminated area and dispose of the respirator. Before each use of the product, please inspect for any rips, damage, tears, etc. These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.

N95 respirators with Cool Flow™ valve are designed to provide protection for applications such as welding or soldering where metal fumes may be present.

  • NIOSH approved N95 rating
  • Adjustable M-noseclip
  • Flame resistant per modified ASTM D2859
  • Recommended for use up to 10X the OSHA PEL or applicable occupational exposure limits, whichever is lower
  • For use when welding, torch cutting, soldering, brazing and working with molten metals
  • Cool Flow™ exhalation valve
TRUSTED_SUSTAINABILITY

Specifications

Aerosol Type N - Not Resistant to Oil
Assigned Protection Factor (APF) 10
Certifications/Compliance ASTM D2859
Certifications (NIOSH) NIOSH 42 CFR 84, N95, approval number: TC-84A-0454
Certifications (OSHA) OSHA 1910.134
Color Code White
Design Type Cup/Molded
Disposable Single-use
Filter Type N95
Material Polypropylene, Polyester
Product Type Particulate Welding Respirator
Size Standard
With Exhalation Valve Yes
Filter Efficiency 95%
For Use With (Application) Welding, torch cutting, soldering, brazing and working with molten metals
Latex Free Yes
Material Headstrap Braided Polyisoprene, Cotton/Polyester
Model 8212
Nose Piece Material Aluminum
Nosepiece Type Standard
Nuisance Odor Relief None
Quantity 80/Cs.
Sterility Non-sterile
With Faceseal Yes
Strap Attachment Type Adjustable Buckle Straps
FDA Cleared Surgical Mask No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.