Learn More
Poltex N95 Respiratory Hygiene Station-Locking
SDP
Catalog No. 22000506
Change view
Click to view available options
Mounting Type:
Countertop
Stand
Wall
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting Type
22-000-506 Wall
22-000-504 Stand
22-000-505 Countertop
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Consumables shown for demonstration purposes only and are not included with the product.
Catalog No. 22-000-506 Supplier Poltex Supplier No. N95HYGSTATW
Add to Cart
Add to Cart
Consumables shown for demonstration purposes only and are not included with the product.
Poltex™ N95 Locking Respiratory Hygiene Station holds one box of N95 respirator masks, one large box of tissue or two half-size boxes, one bottle of hand sanitizer, and one 8.5 x 11 in. sign
  • Lid closes and locks to prevent theft
  • holds standard cone masks when N95 are not needed.
  • Users print their own sign
  • Keys are all keyed the same.
  • Durable 1/4in thick PETG construction
  • Comes with drip tray
  • See image for dimensions.
  • Choose from:
    • Free-standing: Comes with 1/2 in. thick acrylic stand with non-slip rubber bumpons.
    • Wall mount: Sign holder mounts to wall, then station mounts to sign holder.
    • Countertop placement: Comes with 1/2 in. thick acrylic countertop base with non-slip rubber bumpons.
Specifications
Product Type N95 Hygiene Station
For Use With (Equipment) General hygiene products
Mounting Type Wall
Material PETG