Learn More
Cayman Chemical Nα-Benzoyl-DL-Arginine-pNA (hydrochloride), Purity: ≥98%, CAS: 911-77-3, 10 g
SDP
Catalog No. 502947480
Encompass_Preferred
A colorimetric serine protease substrate; release of pNA can be quantified by colorimetric detection at 405 nm as a measure of serine protease activity
Catalog No. 50-294-7480 Supplier Cayman Chemical Supplier No. 3393410 G
A colorimetric serine protease substrate; release of pNA can be quantified by colorimetric detection at 405 nm as a measure of serine protease activity