Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotang Inc Sodium Chloride (0.9%), autoclaved, 250 ml

Catalog No. 50155863 Shop All BIOTANG Inc Products
Encompass

Sodium Chloride (0.9%), autoclaved, 250 ml.

Catalog No. 50-155-863 Supplier Biotang Inc Supplier No. BTBB541
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Chloride (0.9%), autoclaved, 250 ml.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.