Learn More
Aniara Diagnostica LLC Buffered Sodium Citrate 3.2%, 100mL/pouch, 10 pouches
Catalog No. 501008825
Encompass
Medicago Buffered Sodium Citrate 3.2% supplied as preweighed powder mix in sealed pouch yields 100 mL of 3.2% (0.109 M) buffered sodium citrate solution. Use as anti-coagulant for blood storage. 10 pouches.
Catalog No. 50-100-8825 Supplier Aniara Diagnostica LLC Supplier No. A12848010
Medicago Buffered Sodium Citrate 3.2% supplied as preweighed powder mix in sealed pouch yields 100 mL of 3.2% (0.109 M) buffered sodium citrate solution. Use as anti-coagulant for blood storage. 10 pouches.