Learn More
Medchemexpress LLC HY-B1610 1g Medchemexpress, Sodium citrate dihydrate CAS:6132-04-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB16101G
Encompass_Preferred
Medchemexpress, HY-B1610 1g Sodium citrate dihydrate CAS:6132-04-3 Sodium citrate dehydrate is an anticoagulant and also used as a buffer and food preservatives. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2977