Learn More
Medchemexpress LLC HY-B2226 1g Medchemexpress, Sodium copper chlorophyllin B CAS:28302-36-5 Purity:>98%
SDP
Catalog No. 501933116
Encompass_Preferred
Medchemexpress, HY-B2226 1g Sodium copper chlorophyllin B CAS:28302-36-5 Sodium copper chlorophyllin B exerts antiviral activities against Influenza virus and HIV with IC 50 s of 50 to 100 μM for both of them. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3116 Supplier Medchemexpress LLC Supplier No. HYB22261G
Medchemexpress, HY-B2226 1g Sodium copper chlorophyllin B CAS:28302-36-5 Sodium copper chlorophyllin B exerts antiviral activities against Influenza virus and HIV with IC 50 s of 50 to 100 μM for both of them. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.