Learn More
Hach Company Sodium Hypochlorite 0.315% V/V, 500 mL - Hach (52011)
Supplier:  Hach Company 52011
Encompass
Sodium Hypochlorite 0.315% V/V, 500 mL is a prepared laboratory reagent supplied by Hach for total Kjeldahl nitrogen (TKN) analysis using flow injection analysis (FIA). The reagent provides a consistent hypochlorite concentration for automated FIA workflows and routine analytical testing.
  • Prepared reagent for total Kjeldahl nitrogen (TKN) analysis by flow injection analysis (FIA).
  • 0.315% V/V sodium hypochlorite concentration.
  • 500 mL bottle volume.
  • Formulated for use with automated FIA laboratory systems.
  • Consult the manufacturer's safety data sheet for handling, storage, and disposal instructions.
Catalog No. NC2112244