Learn More
Medchemexpress LLC HY-101109 10mg Medchemexpress, Sodium ionophore III CAS:81686-22-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10110910MG
Encompass_Preferred
Medchemexpress, HY-101109 10mg Sodium ionophore III CAS:81686-22-8 Sodium ionophore III (ETH2120) is a Na + ionophore suitable for the assay of sodium activity in blood, plasma, serum. etc. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2043