Learn More
Bioss Na/K ATPase 3 - Extracellul
SDP
Supplier:  Bioss BSM70560M
Encompass_Preferred
Na /K ATPase 3 (Extracellular) Primary Antibody Unconjugated Host Species Mouse Target Species Human Clonality Monoclonal Applications WB IP M016Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-65667