Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Na,K-ATPase Alpha1 Monoclonal Antibody, Rabbit, BSA and Azide Free, 100 μg
SDP

Catalog No. 502729426
Encompass

The Na,K-ATPase α1 (D4Y7E) Rabbit Monoclonal Antibody specifically recognizes endogenous levels of total Na,K-ATPase α1 protein. This recombinant antibody is produced by immunizing animals with a synthetic peptide corresponding to residues surrounding Pro12 of human Na,K-ATPase α1 protein. The formulation is BSA and azide free, making it ideal for custom labeling applications such as fluorophores, metals, lanthanides, and oligonucleotides.

  • Host species: Rabbit
  • Isotype: IgG
  • Molecular weight: 100 kDa
  • Species reactivity: Human and monkey
  • Validated applications: Western blotting, immunohistochemistry (paraffin), and immunofluorescence (immunocytochemistry)
  • Concentration: 1 mg/mL
  • Formulation: 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl; pH 7.8)

Catalog No. 50-272-9426 Supplier Cell Signaling Technology Supplier No. 99935SF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Na,K-ATPase α1 (D4Y7E) Rabbit Monoclonal Antibody specifically recognizes endogenous levels of total Na,K-ATPase α1 protein. This recombinant antibody is produced by immunizing animals with a synthetic peptide corresponding to residues surrounding Pro12 of human Na,K-ATPase α1 protein. The formulation is BSA and azide free, making it ideal for custom labeling applications such as fluorophores, metals, lanthanides, and oligonucleotides.

  • Host species: Rabbit
  • Isotype: IgG
  • Molecular weight: 100 kDa
  • Species reactivity: Human and monkey
  • Validated applications: Western blotting, immunohistochemistry (paraffin), and immunofluorescence (immunocytochemistry)
  • Concentration: 1 mg/mL
  • Formulation: 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl; pH 7.8)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.