Learn More
Hach Company Hach Sodium Nitroprusside Reagent, 1 L - For Ammonia Analysis (FIA)
Catalog No. NC2112246 Shop All Hach Company Products
Encompass
Sodium nitroprusside reagent supplied as a ready-to-use 1 L bottle for quantitative ammonia determination by flow injection analysis (FIA). The reagent is prepared for laboratory use with preserved or unpreserved samples and is intended for use with automated FIA analyzers. Safety Data Sheet (SDS) is available from the manufacturer.
  • Ready-to-use 1 L reagent bottle.
  • Formulated for ammonia determination by flow injection analysis (FIA).
  • Suitable for preserved or unpreserved samples.
  • Compatible with automated FIA analyzers.
  • SDS available from manufacturer; consult SDS before use.
Catalog No. NC2112246 Supplier Hach Company Supplier No. 52006
Sodium nitroprusside reagent supplied as a ready-to-use 1 L bottle for quantitative ammonia determination by flow injection analysis (FIA). The reagent is prepared for laboratory use with preserved or unpreserved samples and is intended for use with automated FIA analyzers. Safety Data Sheet (SDS) is available from the manufacturer.
  • Ready-to-use 1 L reagent bottle.
  • Formulated for ammonia determination by flow injection analysis (FIA).
  • Suitable for preserved or unpreserved samples.
  • Compatible with automated FIA analyzers.
  • SDS available from manufacturer; consult SDS before use.