Learn More
Medchemexpress LLC NA/Neuraminidase H1 150U
SDP
Catalog No. 5000179564
Encompass_Preferred
The NA/Neuraminidase Protein is an influenza virus surface enzyme that cleaves sialic acid residues Inhibiting NA/Neuraminidase Protein is crucial for antiviral drugs Targeting NA/Neuraminidase Protein can help prevent viral release reduce viral spread and potentially treat influenza infections NA/Neuraminidase Protein H1N1 (P03468 HEK293) is the recombinant Virus-derived NA/Neuraminidase protein expressed by HEK293 with tag free This product is an active extract not a recombinant NA protein It originates from cell lysates collected after expressing a DNA sequence encoding the Influenza A virus neuraminidase
Catalog No. 50-001-79564 Supplier Medchemexpress LLC Supplier No. HYP73778150U
The NA/Neuraminidase Protein is an influenza virus surface enzyme that cleaves sialic acid residues Inhibiting NA/Neuraminidase Protein is crucial for antiviral drugs Targeting NA/Neuraminidase Protein can help prevent viral release reduce viral spread and potentially treat influenza infections NA/Neuraminidase Protein H1N1 (P03468 HEK293) is the recombinant Virus-derived NA/Neuraminidase protein expressed by HEK293 with tag free This product is an active extract not a recombinant NA protein It originates from cell lysates collected after expressing a DNA sequence encoding the Influenza A virus neuraminidase