Learn More
Medchemexpress LLC NA/Neuraminidase H1 50ug
SDP
Catalog No. 5000179119
Encompass_Preferred
NA (neuraminidase) proteins play a key role in viral transmission by catalyzing the removal of terminal sialic acid residues from viral and cellular glycoconjugates NA plays a role in determining host range limiting replication and virulence NA is associated with the development and progression of type 2 diabetes mellitus (T2D) NA/Neuraminidase Protein H1N1 (AYV62750 HEK293 His) is the recombinant Virus-derived NA/Neuraminidase protein expressed by HEK293 with N-His labeled tag
Catalog No. 50-001-79119 Supplier Medchemexpress LLC Supplier No. HYP7378450UG
NA (neuraminidase) proteins play a key role in viral transmission by catalyzing the removal of terminal sialic acid residues from viral and cellular glycoconjugates NA plays a role in determining host range limiting replication and virulence NA is associated with the development and progression of type 2 diabetes mellitus (T2D) NA/Neuraminidase Protein H1N1 (AYV62750 HEK293 His) is the recombinant Virus-derived NA/Neuraminidase protein expressed by HEK293 with N-His labeled tag