Learn More
Medchemexpress LLC NA/Neuraminidase In 200U
SDP
Supplier:  Medchemexpress LLC HYP73231200U
Encompass_Preferred
NA/Neuraminidase Protein is a receptor-destroying enzyme that cleaves terminal sialic acids from cellular receptors potentially facilitating viral invasion of the upper airways by targeting sialic acid moieties on airway epithelial cell mucin NA plays a pivotal role in viral propagation by catalyzing the removal of terminal sialic acid residues from both viral and cellular glycoconjugates Moreover the sialidase activity in late endosome/lysosome traffic appears to enhance virus replication This product is an active extract not a recombinant NA protein It originates from cell lysates collected after expressing a DNA sequence encoding the Influenza A virus neuraminidase
Catalog No. 50-001-80280