Learn More
Medchemexpress LLC NA/Neuraminidase In 300U
SDP
Catalog No. 5000181280
Encompass_Preferred
Neuraminidase (NA) is described as a receptor-destroying enzyme because it cleaves a terminal sialic acid from the cellular receptors Whereby NA facilitates viral invasion of the upper airways facilitates virus release during virus budding prevents self-aggregation and ensures the efficient spread of the progeny virus from cell to cell and also plays a role in the determination of host range restriction on replication and virulence NA/Neuraminidase Protein Influenza A H5N1 (H255Y HEK293) is the recombinant Virus-derived NA/Neuraminidase protein expressed by HEK293 with tag free and H255Y mutation It originates from cell lysates collected after expressing a DNA sequence encoding the Influenza A virus neuraminidase
Catalog No. 50-001-81280 Supplier Medchemexpress LLC Supplier No. HYP73239300U
Neuraminidase (NA) is described as a receptor-destroying enzyme because it cleaves a terminal sialic acid from the cellular receptors Whereby NA facilitates viral invasion of the upper airways facilitates virus release during virus budding prevents self-aggregation and ensures the efficient spread of the progeny virus from cell to cell and also plays a role in the determination of host range restriction on replication and virulence NA/Neuraminidase Protein Influenza A H5N1 (H255Y HEK293) is the recombinant Virus-derived NA/Neuraminidase protein expressed by HEK293 with tag free and H255Y mutation It originates from cell lysates collected after expressing a DNA sequence encoding the Influenza A virus neuraminidase