Learn More
Biotang Inc Sodium Phosphate Buffer 0.5M, pH 9.0. Mixture of 500 mM sodium phosphate monobasic and sodium phosphate dibasic, pH 9.0 +/- 0.15. Prepared in 18.2 megohms water and filtered through 0.22 micron filter. BB-2332
Catalog No. 50155783 Shop All BIOTANG Inc Products
Encompass_Preferred
Sodium Phosphate Buffer 0.5M, pH 9.0. Mixture of 500 mM sodium phosphate monobasic and sodium phosphate dibasic, pH 9.0 +/- 0.15. Prepared in 18.2 megohms water and filtered through 0.22 micron filter. BB-2332.
Catalog No. 50-155-783 Supplier Biotang Inc Supplier No. BTBB434
Sodium Phosphate Buffer 0.5M, pH 9.0. Mixture of 500 mM sodium phosphate monobasic and sodium phosphate dibasic, pH 9.0 +/- 0.15. Prepared in 18.2 megohms water and filtered through 0.22 micron filter. BB-2332.