Learn More
Biotang Inc Sodium Phosphate Buffer 0.5M, pH 8.5. Mixture of 500 mM sodium phosphate monobasic and sodium phosphate dibasic, pH 8.5 +/- 0.15. Prepared in 18.2 megohms water and filtered through 0.22 micron filter. BB-2330
Catalog No. 50155782 Shop All BIOTANG Inc Products
Encompass_Preferred
Sodium Phosphate Buffer 0.5M, pH 8.5. Mixture of 500 mM sodium phosphate monobasic and sodium phosphate dibasic, pH 8.5 +/- 0.15. Prepared in 18.2 megohms water and filtered through 0.22 micron filter. BB-2330.
Catalog No. 50-155-782 Supplier Biotang Inc Supplier No. BTBB433
Sodium Phosphate Buffer 0.5M, pH 8.5. Mixture of 500 mM sodium phosphate monobasic and sodium phosphate dibasic, pH 8.5 +/- 0.15. Prepared in 18.2 megohms water and filtered through 0.22 micron filter. BB-2330.